Order Entry
Canada
ContactUsLinkComponent
Human Recombinant SLAMF7 (from HEK293), PE Labeled Protein
Human Recombinant SLAMF7 (from HEK293), PE Labeled Protein
  103889-084
 :  
Human Recombinant SLAMF7 (from HEK293), PE Labeled Protein
  103889-084
 :  SL7HP2H325TESTS
 :  

 

  • Species:
    Human
  • Size:
    25 tests
  • Storage conditions:
    –20 °C, aviod light
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.; 1e6 of the anti-SLAMF7 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human SLAMF7, His Tag (Cat. No. SL7-HP2H3) and negative control protein respectively, PE signals was used to evaluate the binding activity (QC tested).
  • Gene ID:
    Q9NQ25-1;57823;AAH27867
  • Protein synonyms:
    SLAMF7,CD319,CS1,CRACC,19A,FOAP-12
  • Protein/peptide name:
    SLAMF7
  • Molecular weight:
    25.6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-084