Human Recombinant LGR5 (from HEK293), Unconjugated
Catalog # 103889-080
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Human R-Spondin 3 immobilized on CM5 Chip can bind Human LGR5, His Tag (Cat. No. LG5-H52H3) with an affinity constant of 9.52 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:O75473-1;8549;NP_003658.1
- Protein synonyms:LGR5, GPR49, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, G-protein coupled receptor HG38, Leucine-rich repeat-containing G-protein coupled receptor 5
- Protein/peptide name:LGR5
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:62.0 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103889-080