Mouse Recombinant CLEC12A (from HEK293), Unconjugated
103888-972
:
- Conjugation:Unconjugated
- Species:Mouse
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q504P2-1
- Protein synonyms:CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371
- Protein/peptide name:CLEC12A
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:25.7 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103888-972