Human Recombinant TREM2 (from HEK293), Unconjugated
Catalog # 103888-940
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Loaded Human TREM2 Protein, Fc Tag (Cat. No. TR2-H5254) on Protein A Biosensor, can bind Human Apolipoprotein E, His Tag (Cat. No. APE-H5246) with an affinity constant of 106 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q9NZC2-1;54209;NP_061838.1
- Protein synonyms:Triggering receptor expressed on myeloid cells 2,TREM2,TREM-2
- Protein/peptide name:TREM2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:43.9 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103888-940