Human Recombinant PD-1 (from HEK293), Unconjugated
Catalog # 103888-926
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:500 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH8.0 with detergent. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q15116-1;5133;NP_005009.2
- Protein synonyms:PDCD1,PD1,CD279,SLEB2
- Protein/peptide name:PD-1
- Purity:>80% as determined by SDS-PAGE.
- Molecular weight:30.7 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH8.0 with detergent. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103888-926