Order Entry
Canada
ContactUsLinkComponent
SARS-CoV-2 Recombinant S1 protein NTD (from HEK293), Unconjugated
SARS-CoV-2 Recombinant S1 protein NTD (from HEK293), Unconjugated
Catalog # 103888-792
CAS Number:  
SARS-CoV-2 Recombinant S1 protein NTD (from HEK293), Unconjugated
Catalog # 103888-792
Supplier Number:  S1DC52H5100UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    SARS-CoV-2
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    QHD43416.1 (A262S)
  • Protein synonyms:
    S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD
  • Protein/peptide name:
    S1 protein NTD
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    34.9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103888-792

Specifications

About this item