Order Entry
Canada
ContactUsLinkComponent
Human Recombinant NKG2C & CD94 (from HEK293), Precision Avi
Human Recombinant NKG2C & CD94 (from HEK293), Precision Avi
Catalog # 103888-688
CAS Number:  
Human Recombinant NKG2C & CD94 (from HEK293), Precision Avi
Catalog # 103888-688
Supplier Number:  NC4H82E3200UG
CAS Number:  

Specifications

  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; HLA-E*01:03 HLA-A leader3-11 Tetramer-VMAPRTLVL-PE immobilized on CM5 Chip can bind Human NKG2C&CD94 Protein, His,Avitag™ (Cat. No. NC4-H82E3) with an affinity constant of 63.7 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
  • Gene ID:
    P26717-1 (NKG2C) & Q13241-1 (CD94)
  • Protein/peptide name:
    NKG2C & CD94
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    34.3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103888-688

Specifications

About this item