Human Recombinant DR3 / TNFRSF25 (from HEK293), Unconjugated
103888-604
:
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q93038-1;8718;NP_003781.1
- Protein synonyms:TNFRSF25,DR3,APO3,DDR3,TNFRSF12,WSL,WSL1
- Protein/peptide name:DR3 / TNFRSF25
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:20.9 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103888-604