SARS-CoV-2 Recombinant S protein (from HEK293), Precision Avi
Catalog # 103888-442
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Species:SARS-CoV-2
- Size:200 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
- Gene ID:QHD43416.1
- Protein synonyms:Spike,S protein,Spike glycoprotein,S glycoprotein
- Protein/peptide name:S protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:139.8 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103888-442