Human Recombinant Her2 (from HEK293), PE Labeled Protein
103888-128
:
- Species:Human
- Size:25 tests
- Storage conditions:–20 °C, aviod light
- Biological activity:Please contact us for detailed information.
- Gene ID:P04626-1;2064;NP_004439.2
- Protein synonyms:ERBB2,CD340,HER-2,neu,HER2,MLN19,NEU,NGL,TKR1
- Protein/peptide name:Her2
- Molecular weight:73.0 kDa
- Endotoxin level:Low
- Formulation:Please contact us for detailed information.
- Shipping temperature:RT
- Cat. no.:103888-128