Eisco LabGlass® Crow Receivers, Class B
: Wards
High quality glass crow reciever useful for oil distilation and tar tests.
- Made of high quality borosilicate glass 3.3
- Features round base and spout
- Chemical resistant blue graduations
- Class B
- Autoclavable
- Superior chemical durability
- Low temperature gradient - can withstand high temperatures and thermal shocks
- Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Frequently Bought Together