Eisco® Joly Photometer
Joly photometer
Catalog # 470332-232
Supplier: Wards
Supplier #: PH0652G
Jump to:
Product Details & Documents
Used to demonstrate the inverse square law.
- Self standing
- Large base for stability
- Use to demonstrate inverse square law
Specifications
- Description:Joly photometer