Order Entry
Canada
Orders LinkContactUsLinkComponent
Rapid Response™ Strep A Antigen Test Strip, BTNX

Rapid Response™ Strep A Antigen Test Strip, BTNX

Strep-A test strip
Catalog # 76473-508
Rapid Response™ Strep A Antigen Test Strip, BTNX
Catalog # 76473-508
Supplier:  
CAS Number:  
Rapid Response™ Strep A Antigen Test Strip, BTNX
Rapid Response™ Strep A Antigen Test Strip, BTNX

Strep-A test strip

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Rapid Response® Strep A test strip is a rapid, qualitative test for the detection of strep A antigen from throat swab specimens, to aid in the diagnosis of group A streptococcal infection.

  • Sample: Throat swab
  • Format: Strip
  • Specificity: Strep A antigen
  • Time to result: 5 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Lateral flow immunoassay

Delivery information: Includes test strips, extraction test tubes, workstation, sterile swabs, strep a reagents A and B, positive and negative controls, product insert.

Caution: This test is intended for in vitro diagnostic use only.

Specifications

Specifications

  • Description:
    Strep-A test strip

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.