Orders LinkContactUsLinkComponent
ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
Certificates
ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ACLAR® is made from fluorinated-chlorinated resins that provide an exceptional moisture barrier. ACLAR® is crystal clear, biochemically inert, highly resistant to most chemicals and sterilizable by heat or radiation. ACLAR® is used widely in pharmaceutical, medical, sensitive electronics and military packaging.
  • Crystal clear, high UV transparency- ideal for use in UV curing of embedding resin in microscopy
  • Chemically stable and biochemically inert- the product of choice for growing cell cultures
  • Low dielectric constant, high electric strength and dissipation factor– offers excellent cell attachment even through lengthy processing procedures
  • Low surface energy–and separates easily from epoxy
  • Exhibits no detectable autofluorescence– ideal for fluorescent microscopy
  • A non-flammable, non-aging co-polymer– which is suited for sterilization by heat or UV
  • Crystalline melting point is 206 °C (403 °F)– stable in the SEM
  • Flexible and soft– can be sectioned without damage to the ultramicrotome knives
There are two film types–the homopolymer ACLAR® Rx Series and the copolymers ACLAR® 33C.
ACLAR® 33C is a copolymer film consisting primarily of chlorotrifluoroethylene (CTFE). It offers an outstanding moisture barrier, excellent chemical resistance and minimal dimensional change (±2%), making it the best choice for use in microscopy.
Aclar® UltRx 2000 is a 2.00 mil (51 micron) PCTFE homopolymer, high performance barrier film for the pharmaceutical and medical markets. It thermoforms well on conventional blister packaging equipment and provides the best barrier of any clear film.
Product Family Options
Product Information
  • Availability
    Price
  • Log in for availability
    Log in for availability
    Each
    Each - $668.84
    $668.84
    Each
       $668.84
    Supplier Number: 50426-10
  • Log in for availability
    Log in for availability
    Each
    Each - $1,508.19
    $1,508.19
    Each
       $1,508.19
    Supplier Number: 50426-25
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.