Letheen Agar, Modified, Hardy Diagnostics
Catalog #: 76436-466
Supplier: Hardy Diagnostics
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:Glass bottle
- Packaging:6 packs of 12
- Description:Letheen agar, modified
- Size:400 ml fill
- Cat. no.:76436-466
- Fill Volume:16 oz.
Specifications
About this item
Letheen Agar is used for evaluating the bactericidal activity of quaternary ammonium compounds.
- Crystal clear glass construction provides complete visibility
- Features a streamlined design with no handle, making it easier to store and clean
- Easy-to-remove, perforated, shrink wrap ensures product is leak-proof
Customers Who Bought This Also Bought