Order Entry
Canada
ContactUsLinkComponent
Letheen Broth, Modified, Hardy Diagnostics
Letheen Broth, Modified, Hardy Diagnostics
Catalog # 76436-464
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Letheen Broth, Modified, Hardy Diagnostics
Catalog # 76436-464
Supplier:  Hardy Diagnostics
Supplier Number:  U415
CAS Number:  

Specifications

  • Pack type:
    PP bottle, 500 ml
  • Description:
    Letheen broth, modified
  • Size:
    10/pack
  • Cat. no.:
    76436-464
  • Fill Volume:
    250 ml

Specifications

About this item

Letheen Broth, Modified, is for use in microbial recovery from personal care products and for the neutralization of quaternary ammonium compounds in the testing of germicidal activity.

  • Heavy duty bottles are tough and sturdy to reduce breakage
  • Easy-to-remove, perforated, shrink wrap ensures product is leak-proof
  • Optically clear bottles are made of clarified polycarbonate for clear viewing
  • Robust construction of the bottles for excellent impact and chemical resistance