Order Entry
Canada
ContactUsLinkComponent
Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, Hardy Diagnostics
Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, Hardy Diagnostics
  76436-460
 :  
undefined
Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, Hardy Diagnostics
  76436-460
 :  U235
 :  

 

  • Pack type:
    HDPE bottle, 125 ml
  • Description:
    Phosphate buffered saline (PBS) with 0.05% Tween® 20
  • Size:
    12/pack
  • Cat. no.:
    76436-460
  • Fill Volume:
    100 ml

 

 

Used during specimen processing for recovery of bacterial endospores.

Recommended as a component of anthrax screening programs for use in the nasal swab heat-shock procedure to recover Bacillus anthracis endospores.