ContactUsLinkComponent
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
Catalog # 76468-304
undefined
EMS Disc Punch for TEM Preparation, Electron Microscopy Sciences
Catalog # 76468-304
Supplier Number:  50125-10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Wide base specimen punch
  • Cat. no.:
    76468-304
Specifications
About this item
The EMS 310 Disc Punch is designed with 3 mm disc to prepare ductile metals and soft materials for TEM without mechanical distortion. Its solid construction and high tolerance design allows a maximum force to be used with difficult materials while maintaining specimen quality.
  • Preparation of TEM samples without distortion
  • Alignment fixture included for maximum yield and precise specimen location
  • Solid construction for long term use without loss of precision
A specimen alignment tool is included which guarantees maximum specimen yield and precise specimen location. After the disc is punched it is automatically selected from the die and collected in an easily removable tray.
Dimensions - 8 W×8 H×2.5 D"
Weight - 5 lbs.