Orders LinkContactUsLinkComponent
Small Pliers, Excelta
Small Pliers, Excelta
Supplier:   Excelta
Small Pliers, Excelta
Supplier:  Excelta
Certificates
Small Pliers, Excelta
Small Pliers, Excelta
4 Options Available Below
Product Details & Documents
Three star quality small pliers made of carbon steel with flame hardened tips and feature dual leaf springs for smooth action.The grips are soft cushioned foam and are ESD safe.
Product Family Options
Product Information
  • Size
    Length
    Tip size
    Tip style
    Availability
    Price
  • 0.355 W×0.235 D"
    4½"
    0.020 W×0.078 D"
    Flat nose
    Log in for availability
    Log in for availability
    Each
    Each - $153.24
    $153.24
    Each
       $153.24
    Supplier Number: 42I
  • 0.355 W×0.235 D"
    4½"
    Ø 0.020"
    Round nose
    Log in for availability
    Log in for availability
    Each
    Each - $153.24
    $153.24
    Each
       $153.24
    Supplier Number: 43I
  • 0.355 W×0.235 D"
    4½"
    0.020 W×0.039 D"
    Chain nose
    Log in for availability
    Log in for availability
    Each
    Each - $153.24
    $153.24
    Each
       $153.24
    Supplier Number: 44I
  • 0.340 W×0.235 D"
    4½"
    0.020 W×0.035 D"
    Needle nose
    Log in for availability
    Log in for availability
    Each
    Each - $153.24
    $153.24
    Each
       $153.24
    Supplier Number: 47I