ContactUsLinkComponent
1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
Catalog # CA76339-756
CAS Number:  
undefined
1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
Catalog # CA76339-756
Supplier Number:  N0559S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    1 kb Plus DNA ladder for safe stains
  • Size:
    1.25 ml
  • Cat. no.:
    CA76339-756
Specifications
About this item
The 1 kb Plus DNA ladder for safe stains is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye. It has been optimized for use on GelRed, GelGreen and SYBR Safe precast agarose gels.
  • Optimized for visualization on GelRed, GelGreen and SYBR safe precast agarose gels
  • Convenient, ready-to-load format
  • Size range: 100 bp to 10 kb (100 bp and 1 kb ladder in one!)
  • Easy to identify reference bands
This ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.