NEBuffer Set (EcoRI, DpnII), New England Biolabs
Catalog # CA76285-182
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBuffer set(EcoRI, DpnII)
- Size:1.25 ml of each
- Cat. no.:CA76285-182
Specifications
About this item
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity.
Set contains two 1.25 ml vials of each unique NEBuffer for the enzymes EcoRI and DpnII. Most of our enzymes are supplied with one of four standard NEBuffers. Occasionally, an enzyme has specific buffer requirements not met by one of the four standard NEBuffers, in which case the enzyme is supplied with its own unique NEBuffer. 1X NEBuffer EcoRI 100 mM Tris-HCl 50 mM NaCl 10 mM MgCl₂ 0.025% Triton X-100 (pH 7.5 @ 25C) 1X NEBuffer DpnII 50 mM Bis-Tris-HCl 100 mM NaCl 10 mM MgCl2 1 mM DTT (pH 6 @ 25C).