Orders LinkContactUsLinkComponent
NEBNext® Adaptor Dilution Buffer
NEBNext® Adaptor Dilution Buffer
NEBNext adaptor dilution buffer, 9.6 ml
Catalog # CA76326-062
New Product
Supplier #:  B1430S
NEBNext® Adaptor Dilution Buffer
Catalog # CA76326-062
New Product
CAS Number:  
NEBNext® Adaptor Dilution Buffer
NEBNext® Adaptor Dilution Buffer
NEBNext adaptor dilution buffer, 9.6 ml
CAS Number:  
Price
63,00 $
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The NEBNext adaptor dilution buffer is optimized for dilution of the NEBNext adaptor to the appropriate concentration range for efficient ligation to end-repaired, dA-tailed DNA.
  • Can be used to supplement the buffer volume supplied in some NEBNext library prep kits
Specifications
Specifications
  • Description:
    NEBNext adaptor dilution buffer
  • Size:
    9.6 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)