ContactUsLinkComponent
Floor Marking Tape, Blank, Yellow, Accuform
  76471-364
undefined
Floor Marking Tape, Blank, Yellow, Accuform
  76471-364
 :  PTM623YL
  • Width:
    2"
  • Color:
    Yellow
  • Description:
    Floor marking tape, blank
  • Length:
    108'
  • Environmentally Preferable:
  • Cat. no.:
    76471-364
Marking tape is used to mark specific areas and promote safety by identifying potential hazard areas.
  • Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
  • Permanent self-adhesive back with no liner
Extremely durable floor marking tape is waterproof and wear-resistant, while offering minor resistance to acids, oils, grease, caustics, alkalis, and most solvents.