Barricade Tape, Blank, Accuform
Catalog #: 76471-360
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Length:1000'
- Width:3"
- Color:Yellow
- Description:Barricade tape, blank
Specifications
About this item
Barricade tape temporarily sections-off hazards and danger zones quickly and effectively.
- Easily cut and secure with adhesive tape, nail, staples, or tie-off
Lightweight polyethylene plastic material is durable for indoor and outdoor environments.
Customers Who Bought This Also Bought