3-Biotin-GTP , New England Biolabs
Catalog # 103501-478
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:3-Biotin-GTP 5 mM
- Size:0.5 mol
- Cat. no.:103501-478
Specifications
About this item
Supplied in Ultrapure water as a Triethylammonium salt at pH 7.0
Molecular formula: C₃₁H₅₀N₁₁O₁₉P₃S
Molecular weight: 1005.78 g/mol
Extinction coefficient: 260 = ~11700 Lmol⁻¹ cm⁻¹