NEBNext Multiplex Oligos for Illumina (Dual Index Primers Set 2), New England Biolabs
CA76284-672
:
- Description:NEBNext Multiplex Oligos for Illumina (dual index primers set 2)
- Cat. no.:CA76284-672
- No. of Reactions:96
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification.
- High efficiency adaptor ligation
- Minimized adaptor-dimer formationIncludes index primers for library multiplexing
- Increased sample identification specificityIncludes USER Enzyme
: This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing.