The NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina uses a template switching method to generate full length cDNAs directly from single cells or 2 pg to 200 ng RNA, followed by conversion to sequence-ready libraries using the Ultra II FS workflow.
- Experience unmatched detection of low abundance transcripts
- Rely on consistent transcript detection for a wide range of input amounts and sample types
- Observe uniform transcript coverage, regardless of input amount or sample type
- Use with a variety of RNA inputs, including cultured or primary cells, or total RNA
This unique workflow enables generation of the highest yields from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Environmentally Preferable: