ContactUsLinkComponent
NEB Cloning Competent E. coli Sampler, New England Biolabs
NEB Cloning Competent E. coli Sampler, New England Biolabs
Catalog # CA76299-644
CAS Number:  
undefined
NEB Cloning Competent E. coli Sampler, New England Biolabs
Catalog # CA76299-644
Supplier Number:  C1010S
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    NEB cloning competent E. coli sampler
  • Size:
    8 vials×0.05 ml
  • Cat. no.:
    CA76299-644
Specifications
About this item
The NEB Cloning Competent E.coli Sampler allows to sample four of popular chemically competent strains.
  • NEB 5-alpha: strain for basic cloning; Derivative of the DH5
  • NEB 10-beta: derivative of the DH10B
  • NEB Stable: Ideal for isolation of plasmid clones containing repeat elements and unstable inserts
  • NEB Turbo: Features fast colony growth (6.5 hours) and tight expression control (lacIq)
The Sampler includes two 0.05 ml tubes of each of the following strains (as well as outgrowth medium and control).