Specifications
- Description:NEB 10-beta/stable outgrowth medium
- Size:100 (4×25 ml)
- Cat. no.:76196-414
Specifications
About this item
Outgrowth medium recommended for use with NEB 10-beta or NEB Stable Competent E. coli.
- Use to dilute the outgrowth prior to plating
Add NEB 10-beta/Stable Outgrowth Medium to freshly transformed NEB 10-beta or NEB Stable E. coli cells and incubate for up to 1 hour before plating on selective media.