Order Entry
Canada
ContactUsLinkComponent
 
Hughes ™ Eyewash Additive Solution, Justrite®
  CA76471-008
 :  Justrite
undefined
Hughes ™ Eyewash Additive Solution, Justrite®
  CA76471-008
 :  Justrite
 :  ADDR4PKCAN

 

  • Capacity:
    8 oz.
  • Color:
    Green
  • Cat. no.:
    CA76471-008
  • Weight:
    0.65 lbs.

 

 

Eyewash additive preserves water in portable, self-contained eyewash stations to prevent bacterial growth.

  • Water preservative solution for use with self-contained eye/face wash stations
  • Each 8oz. bottle preserves 5 to 20 gal. of water for up to 120 days
  • Sealed bottle has a shelf life of 3 years

Eyewash additive treats water to help avoid additional damage to an already injured eye, while also extending the useful life of eyewash water. Labeled in accordance with USFDA.

Simply fill a clean eyewash station with potable water and add the additive solution. Treated water reduces the frequency of water changes, minimizing the maintenance required to stay compliant (ANSI Z358.1-2014, Section 5.5.3 recommends weekly visual checks of flushing fluid). A tamper-evident seal offers extra security.

For use with Hughes 10GFEW,16GFEW, 15GEW, 28GEW, and 38GEW and other portable or self-contained eyewashes.