To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Tube revolver
- Cat. no.:76421-798
- Rotation Speed:10 - 40 rpm
- Operating Temperature:4...60 °C
Specifications
About this item
Using a combination of up to four paddle the tube revolver can hold a combination of paddles.
- Maintenance-free motor with quiet operation
- Plug and Play operation
- Adjustable paddle angle
- Includes five interchangeable rotisseries
With seven 10/15 ml tubes, seven 5/7 ml tubes, fifteen 1.5/2.0 ml tubes, or twenty one 0.5/0.8 ml tubes. Or one paddle of six 50 ml tubes.
Max. power: 10 W