Monarch® RBC Lysis Buffer, New England Biolabs
Catalog # CA76470-768
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:Monarch® RBC lysis buffer
- Size:160 ml
- Cat. no.:CA76470-768
Specifications
About this item
The Monarch® RBC lysis buffer is specifically designed to lyse red blood cells (erythrocytes) while leaving leukocytes intact. The separate RBC lysis step enables removal of hemoglobin to increase the purity of the blood prep. The RBC Lysis Buffer also removes nucleases to enable the subsequent purification of HMW DNA into the megabase (Mb) range in size.
- A component of the Monarch HMW DNA extraction kit for cells and blood
Store this buffer at 4 °C.