Electrode Filling Solution, 3 M KCl Electrolyte
Catalog # 76359-830
Supplier: Mettler Toledo
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Bottle
- Description:Potassium chloride solution 3 M KCl
- Size:250 ml
- Environmentally Preferable:
- Cat. no.:76359-830
- Ion:Barium, fluoride, calcium, sodium, lithium
Specifications
About this item
Mettler-Toledo offers electrolytes for every application. Fill-up your electrode with correct and fresh electrolyte for a precise measurement.
Sensor used: Reference cell, combined electrode
Parameter: pH and ISE