ContactUsLinkComponent
TaqDog Premium DNA Polymerase with 10X Reaction Buffers (no dNTP), Bulldog Bio
TaqDog Premium DNA Polymerase with 10X Reaction Buffers (no dNTP), Bulldog Bio
  103881-410
 :  
undefined
TaqDog Premium DNA Polymerase with 10X Reaction Buffers (no dNTP), Bulldog Bio
  103881-410
 :  TQDG500
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    TaqDog Premium DNA polymerase with 10X reaction buffer (no dNTP)
  • Size:
    50000 units
  • Cat. no.:
    103881-410
TaqDog PCR enzymes can be used in many types of experiments that require a robust and active Taq polymerase. It’s highly stable, and able to be diluted, making it perfect for labs that are looking for a reliable and cost effective PCR enzyme.
TaqDog Premium is a great choice for everyday PCR needs. This recombinant DNA polymerase is derived from Thermus aquaticus is perfect for 3-step, 2-step, and touchdown protocols, as well as completely compatible with common SYBR™ Green-based qPCR buffers. Each kit includes 10X reaction buffer and is optimized for improved amplification efficiency. Better efficiency allows many protocols to be completed in under 1 hour — that is if your thermal cycler can keep up!
TaqDog Premium is also a great enzyme to consider for dramatically reducing the cost of qPCR experiments. Starting at only a few pennies per unit, its ideal for labs performing mouse tail genotyping, and other high volume PCR processing. The purity of TaqDog Premium is so good, that it’s still 100% fully active after 14 days at 37 °C. That’s why we ship it at room temperature — a cost savings for your lab and waste savings for the environment.
Use for Standard PCR up to 5 kb, Hot start PCR, Mouse tail genotyping, Sybr Green qPCR and Bisulfite PCR.