Order Entry
Canada
ContactUsLinkComponent
 
Pliers, Forms J-Hook, Excelta
  75880-624
 :  Excelta
undefined
Pliers, Forms J-Hook, Excelta
  75880-624
 :  Excelta
 :  554J-US

 

  • Description:
    Pliers, misc. former, straight 5 star quality
  • Length:
    150 mm (6")
  • Material:
    Carbon
  • Cat. no.:
    75880-624
  • Tip Size:
    0.250"
  • Supplier No.:
    554J-US

 

 

Designed to forms a 'J' hook in leads up to a 0.031" diameter.

  • Ideal for forming 'J Hook' in wire
  • Ergonomic grips, dual leaf springs

The 554J-US five star 6" long forming tool forms a 'J' hook in leads up to a 0.031" diameter with an inside diameter of 0.060" and an outside diameter of 0.125". It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs.