Orders LinkContactUsLinkComponent
Removable Polypropylene Labels, Brady
Supplier:   Brady Worldwide
Removable Polypropylene Labels, Brady
Supplier:  Brady Worldwide
Certificates
Removable Polypropylene Labels, Brady
Removable Polypropylene Labels, Brady
6 Options Available Below
Product Details & Documents
These B-521 thermal transfer labels do not leave any adhesive residue behind when they are removed.
  • Semi-gloss finish
  • Removable acrylic adhesive type
Ideal for use in temporary labeling applications where solvent resistance and print performance are required. Have excellent low temperature resistance. If using this part in an IP series printer or i5100 printer, white print ribbon is not recommended.
 : Not recommended for long-term outdoor use.
Product Family Options
Product Information
  • Material
    Color
    For Use With
    Core size
    Dimensions
    Packaging
    Availability
    Price
  • 76461-920
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    11.1×12.7 mm (0.44×0.5")
    10000 labels/roll, 1 roll (10000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - 252,28 $
    252,28 $
    New Customer Promotional Price
    Roll of 1
       252,28 $
    Supplier Number: THT-12-521-10
  • 76461-912
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    12.7×25.4 mm (0.5×1")
    10000 labels/roll, 1 roll (10000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - 292,58 $
    292,58 $
    New Customer Promotional Price
    Roll of 1
       292,58 $
    Supplier Number: THT-5-521-10
  • 76461-908
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    12.7×25.4 mm (0.5×1")
    20000 labels/roll, 2 rolls (40000 labels)
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - 339,75 $
    339,75 $
    New Customer Promotional Price
    Pack of 2
       339,75 $
    Supplier Number: THT-59-521-10
  • 76461-918
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    25.4×50.8 mm (1×2")
    3000 labels/roll, 1 roll (3000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - 269,94 $
    269,94 $
    New Customer Promotional Price
    Roll of 1
       269,94 $
    Supplier Number: THT-17-521-3
  • 76461-904
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    25.4×76.2 mm (1×3")
    3000 labels/roll, 1 roll (3000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - 366,20 $
    366,20 $
    New Customer Promotional Price
    Roll of 1
       366,20 $
    Supplier Number: THT-18-521-3
  • 76461-910
    B-521 PP-label, removable
    B-521 PP-label, removable
    PP
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    50.8×76.2 mm (2×3")
    1000 labels/roll, 1 roll (1000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - 248,68 $
    248,68 $
    New Customer Promotional Price
    Roll of 1
       248,68 $
    Supplier Number: THT-19-521-1