Orders LinkContactUsLinkComponent
Super Erecta® Post-Type Wall Mount Double Shelf Support
Super Erecta® Post-Type Wall Mount Double Shelf Support
Super Erecta® Post-Type Wall Mount Double Shelf Support
Super Erecta® Post-Type Wall Mount Double Shelf Support
Super Erecta® Post-Type Wall Mount Double Shelf Support
Double shelf support for use with post-type wall mounted shelving.
  • Use double shelf supports to create runs of multiple shelves
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Ordering information: A single shelf support (sold separately) is required at the end of each run.
Product Family Options
  • Description
    Color
    Finish
    Material
    Width
    Length
    Height
    Load capacity
  • Post-type wall mount double shelf support
    Grey
    Metroseal epoxy
    Steel
    2¼"
    16⅝"
    7⅜"
    250 lbs.
    Each
    Each - $161.00
    $161.00
    Each
       $161.00
    Minimum order quantity is 1
     : 2WS14K4
  • Post-type wall mount double shelf support
    Grey
    Metroseal epoxy
    Steel
    2¼"
    20⅝"
    7⅜"
    250 lbs.
    Each
    Each - $168.00
    $168.00
    Each
       $168.00
    Minimum order quantity is 1
     : 2WS18K4
  • Post-type wall mount double shelf support
    Grey
    Metroseal epoxy
    Steel
    2¼"
    26⅝"
    7⅜"
    250 lbs.
    Each
    Each - $177.00
    $177.00
    Each
       $177.00
    Minimum order quantity is 1
     : 2WS24K4