Human Recombinant CD39 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Human CD39 Protein, Mouse IgG2a Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.1 EU per μg by the LAL method.
- Gene ID:P49961-1
- Protein synonyms:CD39|Ecto-ATPDase 1|Ecto-ATPase 1|ENTPD1|Entpd1|NTPDase 1
- Protein/peptide name:CD39
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:77.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 500 mM Argnine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
Specifications
Frequently Bought Together