Human Recombinant CD36 (from HEK293 Cells), Biotin
103871-640
:
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Tag sequence:Precision Avi
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Protein synonyms:GP4|GP3B|FAT|GPIV|gpIV|SCARB3|CHDS7|glycoprotein IIIb|CD36|glycoprotein IV|BDPLT10|gpIIIb|PASIV|Platelet Glycoprotein 4
- Protein/peptide name:CD36
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:50.2 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-640
Biotinylated Human CD36 / SR-B3 Protein, His,Avitag™, ACROBiosystems