Cynomolgus Monkey, Rhesus macaque Recombinant BCMA (from HEK293 cells), Biotin
Catalog # 103871-172
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyRhesus monkey
- Size:200 µg
- Tag sequence:Precision Avi
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:G7Q0I4-1
- Protein synonyms:CD269|TNFRSF17|BCM
- Protein/peptide name:BCMA
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:9.6 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-172
Specifications
About this item
Biotinylated Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His,Avitag™, ACROBiosystems