Order Entry
Canada
ContactUsLinkComponent
Human Recombinant APRIL (from HEK293 cells)
Human Recombinant APRIL (from HEK293 cells)
Catalog # 103871-334
CAS Number:  
Human Recombinant APRIL (from HEK293 cells)
Catalog # 103871-334
Supplier Number:  APLH52D1100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Tag sequence:
    active trimer (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Loaded Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) on HIS1K Biosensor, can bind Human BCMA, Fc Tag (Cat. No. BC7-H5254) with an affinity constant of 1.88 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) on HIS1K Biosensor, can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with an affinity constant of 75 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    O75888-1
  • Protein synonyms:
    APRIL|TNFSF13|TALL2|TRDL-1|ZTNF2|TALL-2|CD256
  • Protein/peptide name:
    APRIL
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    50.2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-334

Specifications

About this item

Human APRIL / TNFSF13 Protein, His,Flag Tag, active trimer (MALS verified), ACROBiosystems