Human Recombinant Alpha-Synuclein (from E. coli)
Catalog # 103814-828
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:200 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P37840-1
- Protein synonyms:PARK1|SNCA|alpha-Synuclein|NACP
- Protein/peptide name:Alpha-Synuclein
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:16.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 100 mM NaAC, pH7.0. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103814-828
Specifications
About this item
Human Alpha-Synuclein Protein, His Tag, ACROBiosystems