Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant 4-1BB (from HEK293 cells), Biotin
Catalog # 103870-916
CAS Number:  
Human Recombinant 4-1BB (from HEK293 cells), Biotin
Catalog # 103870-916
Supplier Number:  41BH82E6200UG
CAS Number:  

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    200 µg
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Biotinylated Human 4-1BB Protein, His,Avitag™;;;;;;;
  • Gene ID:
    Q07011-1
  • Protein synonyms:
    CD137|TNFRSF9|ILA|CDw137|4-1BB
  • Protein/peptide name:
    4-1BB
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    20.9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103870-916

Specifications

About this item

Biotinylated Human 4-1BB / TNFRSF9 Protein, His,Avitag™(MALS verified), ACROBiosystems