Order Entry
Canada
ContactUsLinkComponent
 
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Catalog # 103871-318
CAS Number:  
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Catalog # 103871-318
Supplier Number:  SPDC52H5100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Size:
    100 µg
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Human ACE2, Fc Tag (Cat. No. AC2-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind SARS-CoV-2 S protein RBD (V483A), His Tag (Cat. No. SPD-C52H5) with an affinity constant of 4.13 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Human ACE2, Fc Tag (Cat. No. AC2-H5257) on Protein A Biosensor, can bind SARS-CoV-2 S protein RBD (V483A), His Tag (SPD-C52H5) with an affinity constant of 6.15 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) on AHC Biosensor, can bind SARS-CoV-2 S protein RBD (V483A), His Tag (Cat. No. SPD-C52H5) with an affinity constant of 4.57 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Protein synonyms:
    Spike|S glycoprotein RBD|Spike glycoprotein Receptor-binding domain|Spike protein RBD|S protein RBD|COVID-19
  • Protein/peptide name:
    S protein RBD
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    27.0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-318

Specifications

About this item

SARS-CoV-2 (COVID-19) S protein RBD (V483A), His Tag, ACROBiosystems