Order Entry
Canada
ContactUsLinkComponent
 
SARS-CoV-2 Recombinant S protein (from HEK293 cells), Biotin
  103870-968
 :  
SARS-CoV-2 Recombinant S protein (from HEK293 cells), Biotin
  103870-968
 :  SPNC82E9200UG
 :  

 

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Size:
    200 µg
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein (R683A, R685A, K986P, V987P), His Tag, active trimer (Cat. No. SPN-C52H9) with a linear range of 0.5-16 ng/mL (QC tested).
  • Protein synonyms:
    Spike glycoprotein|Spike, S protein|S glycoprotein|COVID-19
  • Protein/peptide name:
    S protein
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    139.7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103870-968

 

 

Biotinylated SARS-CoV-2 S protein, His,Avitag™, Super stable trimer (MALS verified), ACROBiosystems