Order Entry
Canada
ContactUsLinkComponent
 
Rabbit Recombinant M-CSF R (from HEK293 Cells)
  103815-388
 :  
Rabbit Recombinant M-CSF R (from HEK293 Cells)
  103815-388
 :  CSRR82H5100UG
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Species:
    Rabbit
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    G1TTE0-1
  • Protein synonyms:
    CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
  • Protein/peptide name:
    M-CSF R
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    55.2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103815-388

 

 

Rabbit M-CSF R / CSF1R / CD115 Protein, His Tag, ACROBiosystems