Order Entry
Canada
ContactUsLinkComponent
 
Rabbit Recombinant Complement C5 (from HEK293 Cells)
  103814-978
 :  
Rabbit Recombinant Complement C5 (from HEK293 Cells)
  103814-978
 :  CO5R52H4100UG
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rabbit
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Protein synonyms:
    CPAMD4|Complement C5|C5
  • Protein/peptide name:
    Complement C5
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    73.8 kDa (β chain) & 111.9 kDa (α chain)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103814-978

 

 

Rabbit Complement C5 Protein, His Tag, ACROBiosystems