Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant PSMA (from HEK293 Cells)
Mouse Recombinant PSMA (from HEK293 Cells)
  103815-014
 :  
Mouse Recombinant PSMA (from HEK293 Cells)
  103815-014
 :  PSAM5245100UG
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    O35409-1
  • Protein synonyms:
    FOLH|FOLH1|PSM|GCPII|NAALAD1|PSMA|FGCP|NAALADase I|GIG27
  • Protein/peptide name:
    PSMA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    81.5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in MES and NaCl. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103815-014

 

 

Mouse PSMA / FOLH1 Protein, His Tag (active enzyme), ACROBiosystems