Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant Mesothelin (from HEK293 cells)
Mouse Recombinant Mesothelin (from HEK293 cells)
Catalog # 103871-500
CAS Number:  
Mouse Recombinant Mesothelin (from HEK293 cells)
Catalog # 103871-500
Supplier Number:  MSNM52H3100UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    Q61468-1
  • Protein synonyms:
    MPF|MSLN|Mesothelin
  • Protein/peptide name:
    Mesothelin
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    36.0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-500

Specifications

About this item

Mouse Mesothelin / MSLN (298-600) Protein, His Tag (MALS verified), ACROBiosystems