Mouse Recombinant ICOS (from HEK293 cells)
Catalog # 103815-294
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Human B7-H2, His Tag (Cat. No. B72-H5221) at 1 μg/mL (100 μL/well) can bind Mouse ICOS (C137S, C138S), Fc Tag (Cat. No. ICS-M5259) with a linear range of 0.3-10 ng/mL (QC tested).
- Gene ID:Q9WVS0-1 (C137S, C138S)
- Protein synonyms:Inducible T-cell costimulator|AILIM|CD278|ICOS
- Protein/peptide name:ICOS
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:40.6 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-294
Specifications
About this item
Mouse ICOS / CD278 (C137S, C138S) Protein, Fc Tag, ACROBiosystems